Learn More
Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12179910MG
Encompass_Preferred
Taurohyocholic acid is a taurine-conjugated bile acid derived from hyocholic acid, provided as a white to off-white solid for biochemical and in vitro research applications.
  • High purity suitable for analytical and biological assays.
  • Taurine-conjugated bile acid used as an analytical reference and research reagent.
  • Soluble in DMSO at high concentrations; ultrasonic agitation may be required.
  • Stable as a powder when stored at -20 °C for long-term storage.
  • Supplied in small research pack sizes for assay development and method validation.
Catalog No. 50-002-30240