Learn More
Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 100 MG

Supplier: Medchemexpress LLC HY121799100MG
Taurohyocholic acid is a taurine-conjugated bile acid supplied as a high-purity research reagent and endogenous metabolite reference standard for biochemical and metabolic studies.
- High purity (99.7%).
- Molecular weight 515.70 g/mol.
- Molecular formula C26H45NO7S.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Available in small pack sizes, including 100 MG.
- Store powder at -20°C (3 years) or 4°C (2 years); store in solvent at -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.