Learn More
Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12179925MG
Encompass_Preferred
Taurohyocholic acid is a taurine-conjugated hyocholic bile acid supplied as a white to off-white solid for biochemical research, commonly used as an endogenous metabolite probe and in in vitro studies of bile acid metabolism and signaling.
  • High purity suitable for analytical and biochemical studies.
  • Soluble in DMSO; freshly opened DMSO is recommended for best solubility.
  • Stable as a powder when stored at low temperature for extended periods.
  • Available in multiple pack sizes for flexible experimental needs.
  • White to off-white solid appearance for easy visual inspection.
Catalog No. 50-002-32816