Learn More
Medchemexpress LLC Taurohyocholic acid | 32747-07-2 | 99.7% | 50 MG

Supplier: Medchemexpress LLC HY12179950MG
Taurohyocholic acid is a taurine-conjugated hyocholic bile acid used as a research reagent in biochemical and pharmacological studies. It is supplied as a high-purity powder with documented analytical data and recommended storage and solubility conditions.
- Chemical name: Taurohyocholic acid.
- Cas number: 32747-07-2.
- Chemical formula: C26H45NO7S.
- Molecular weight: 515.70.
- Purity: 99.7% (reported by manufacturer).
- Solubility: DMSO 100 mg/mL; ultrasonic assistance recommended; use freshly opened DMSO.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Applications: endogenous metabolite and bile-acid research; reported as a predictor of response to tyrosine kinase inhibitors combined with PD-1 inhibitors in hepatocellular carcinoma.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.