Learn More
Medchemexpress LLC Taurohyodeoxycholic 100mg | 386523 | 100 MG

Supplier: Medchemexpress LLC HY114360100MG
Taurohyodeoxycholic acid is a taurine-conjugated secondary bile acid used in biochemical and pharmacology research to study bile acid signaling, inflammation, and metabolic pathways. It is supplied for laboratory use in solid form or as a solution and is compatible with common solvents when prepared according to recommended conditions.
- High chemical purity suitable for analytical and pharmacological studies.
- Available in multiple package sizes for small-scale research and larger requests.
- Soluble in DMSO at high concentration; ultrasonic assistance recommended for dissolution.
- Recommended storage conditions preserve compound stability and shelf life.
- Applicable as a standard or reagent in bile acid and metabolic assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.