Learn More
Medchemexpress LLC Taurolidine | 19388-87-5 | 98.9% | 284.363 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW011522R
Taurolidine (Standard) is the analytical standard of Taurolidine. Intended for research and analytical applications, it acts as a potent antimicrobial and anticancer agent. It inhibits cell proliferation, induces apoptosis and autophagy, and can rescue mice from sepsis-associated lethality.
- Molecular formula: C7H16N4O4S2
- Target: Bacterial, apoptosis, antibiotic, autophagy, reference standards
- Pathway: Anti-infection, apoptosis, autophagy, others
- Caution: Product has not been fully validated for medical applications; for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.