Learn More
Medchemexpress LLC Taurolithocholic acid | 516-90-5 | 99.7% | 483.70 g·mol^-1 | C26H45NO5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1133085MG
Encompass_Preferred
Taurolithocholic acid is a taurine-conjugated bile acid used as a research reagent. It functions as a calcium channel agonist and modulates cell viability, S phase entry, and DNA synthesis in RMCCA-1 cells, with reported effects on EGFR and ERK signaling. Batch-specific documentation is provided for analytical verification.
Catalog No. 50-002-63905