Learn More
Cayman Chemical TauromurIcholIc AcId-d4s 1mg
SDP
Supplier:  Cayman Chemical 270401 MG
Encompass_Preferred
An internal standard for the quantification of tauro-β-muricholic acid by GC- or LC-MS
Catalog No. 50-293-7760