Learn More
Medchemexpress LLC Tauroursodeoxycholate-d5 (tauroursodeoxycholic acid-d5) | 1207294-25-4 | 504.73 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY19696S1MG
Encompass_Preferred
Tauroursodeoxycholate-d5 is the deuterium labeled Tauroursodeoxycholate. Tauroursodeoxycholate (Tauroursodeoxycholic acid) is an endoplasmic reticulum (ER) stress inhibitor. It significantly reduces the expression of apoptosis molecules, such as caspase-3 and caspase-12, and also inhibits ERK.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-03264