Learn More
Medchemexpress LLC Tavapadon | 1643489-24-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11948650MG
Encompass_Preferred
Tavapadon is an orally active, brain-penetrant dopamine D1/D5 receptor partial agonist used in preclinical research into Parkinson's disease and dopaminergic signaling. It is supplied as a high-purity research chemical suitable for in vitro and in vivo pharmacology studies.
  • Orally active D1/D5 receptor partial agonist.
  • High purity: 99.94%.
  • Available as solid doses (5-100 mg) and 10 mM solution in DMSO.
  • Molecular formula C19H16F3N3O3; molecular weight 391.34.
  • Intended for pharmacology and preclinical research applications.
Catalog No. 50-002-33172