Learn More
Cayman Chemical -TaxIfolIn 500mg
SDP
Supplier:  Cayman Chemical 23234500 MG
Encompass_Preferred
A flavonol with antioxidant activity; increases cell viability of FeCl2 and H2O2-treated bmMSCs (1-100 UG/ml); scavenges PTIO radicals in a pH-dependent manner (IC50s = 2.6-0.4 mM for pH 5-9, respectively); protects against H2O2- and xanthine/xanthine oxidase-induced oxidative injury in primary cultured rat cortical cells (IC50 = 7.8 UG/ml); inhibits lipid peroxidation in rat brain homogenates (IC50 = 1.02 UG/ml)
Catalog No. 50-298-6815