Learn More
Medchemexpress LLC HY-N0136 50mg Medchemexpress, Taxifolin CAS:480-18-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN013650MG
Encompass_Preferred
Medchemexpress, HY-N0136 50mg Taxifolin CAS:480-18-2 Taxifolin ((+)-Dihydroquercetin) exhibits important anti- tyrosinase activity. Taxifolin exhibits significant inhibitory activity against collagenase with an IC 50 value of 193.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3418