Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taxifolin 7-O-α-L-rhamnoside | 137592-12-2 | 20 MG
SDP

Supplier:  Medchemexpress LLC HYN431020MG

Encompass_Preferred

Taxifolin 7-O-rhamnoside (Taxifolin 7-O-α-L-rhamnoside) is a flavonoid isolated from *Hypericum japonicum*. It is intended for research use only.

  • Purity: 98.13%
  • Appearance: Solid, off-white to light yellow
  • Molecular weight: 450.39
  • Formula: C21H22O11
  • Soluble in DMSO at 125 mg/mL
  • Storage: 4°C, protected from light
  • In solvent, -80°C for 6 months or -20°C for 1 month (protect from light)

Catalog No. 50-004-20921


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.