Learn More
Medchemexpress LLC Taxifolin 7-O-rhamnoside | 137592-12-2 | MFCD04039845 | 99.6% | 450.39 g/mol | C21H22O11 | 10 MG

Supplier: Medchemexpress LLC HYN431010MG
Taxifolin 7-O-rhamnoside is a flavonoid glycoside isolated from Hypericum japonicum and provided as a high-purity research reagent for biochemical and natural product studies. The compound's chemical properties and small pack sizes make it suitable for assay development and exploratory laboratory work.
- Natural flavonoid glycoside isolated from Hypericum japonicum.
- Molecular formula C21H22O11 and molecular weight 450.39 g/mol.
- CAS number 137592-12-2.
- High reported purity (≈99.6%).
- Available in milligram pack sizes suitable for research, including 10 mg.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.