Learn More
ATCC Taylorella asinigenitalis (ATCC 700933) freeze-dried culture, strain UCD-1
Supplier: ATCC 700933
Taylorella asinigenitalis Jang et al. is supplied as a freeze-dried bacterial culture for laboratory research and propagation. The strain corresponds to the UCD-1 isolate and includes recommended recovery conditions for consistent revival and growth.
- Freeze-dried culture for long-term storage.
- Intended for laboratory research and propagation of Taylorella species.
- Recover on CEM medium under microaerophilic conditions.
- Incubate at 37 °C with a 95% air, 5% CO2 atmosphere.
- Supplied with handling and storage instructions to maintain viability.