Learn More
ATCC Taylorella equigenitalis (Taylor et al.) Sugimoto et al. - ATCC 49729, freeze-dried reference strain
Supplier: ATCC 49729
ATCC 49729 is the type strain of Taylorella equigenitalis, a fastidious, Gram-negative coccobacillus and the causative agent of contagious equine metritis (CEM). The strain was isolated from the vaginal discharge of a mare and is supplied in a freeze-dried format for reference and diagnostic microbiology use. Rehydrate and culture following supplier instructions under microaerophilic conditions.
- Type strain for Taylorella equigenitalis.
- Freeze-dried for long-term storage.
- Isolated from vaginal discharge of mare.
- Grows on GC agar/broth; incubate at 37°C in 5% CO2.
- Provided for reference, diagnostic, and comparative studies.