Learn More
Medchemexpress LLC Tazemetostat | 1403254-99-8 | 572.74 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13803R
Encompass_Preferred
Tazemetostat (EPZ-6438) is a potent, selective, and orally available EZH2 inhibitor. It inhibits human polycomb repressive complex 2 (PRC2)-containing wild-type EZH2 with a Ki value of 2.5 nM. It inhibits EZH2 with IC50s of 11 and 16 nM in peptide and nucleosome assays, respectively, and rat EZH2 with an IC50 of 4 nM. Tazemetostat also inhibits EZH1 with an IC50 of 392 nM. It induces apoptosis and differentiation specifically in SMARCB1-deleted MRT cells.
  • Potent, selective, and orally available EZH2 inhibitor.
  • Induces apoptosis and differentiation specifically in SMARCB1-deleted MRT cells.
Catalog No. 50-003-52146