Learn More
Med Vet International Safe-T-Fill Capillary Blood Collection Tube K2 EDTA Additive 125 µL Pierceable Attached Cap Plastic Tube, 076011, 50/Pack
Supplier: Med Vet International 333707PK
Liquid dipotassium EDTA The attached cap is color coded to designate the type of additive that is in both the capillary tube and the micro tube Pre-assembled with a capillary straw, attached pierceable cap, and a microtube Printed on the micro tube are the lot number and expiration date as well as fill lines