Learn More
Med Vet International Safe-T-Fill Capillary Blood Collection Tube K2 EDTA Additive 125 µL Pierceable Attached Cap Plastic Tube, 076011, 500/Case
Supplier:  Med Vet International 333707CS
Encompass_Preferred
Liquid dipotassium EDTA The attached cap is color coded to designate the type of additive that is in both the capillary tube and the micro tube Pre-assembled with a capillary straw, attached pierceable cap, and a microtube Printed on the micro tube are the lot number and expiration date as well as fill lines
Catalog No. 50-304-3240