Learn More
Medchemexpress LLC HY-19750 5mg Medchemexpress, TBA-7371 CAS:1494675-86-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY197505MG
Encompass_Preferred
Medchemexpress, HY-19750 5mg TBA-7371 CAS:1494675-86-3 TBA-7371 is a potent, noncovalent DprE1 inhibitor. TBA-7371 has potent antitubercular activity . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2175