Learn More
Enzo Life Sciences Tick-borne Encephalitis Virus NS1 protein, (recombinan), (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2800100
Encompass_Preferred
Recombinant Tick-borne encephalitis virus NS1 protein produced in HEK293 human cells. Protein contains a C-terminal 6x His-tag. Recombinant Tick-borne encephalitis virus NS1 protein produced in HEK293 human cells. Protein contains a C-terminal 6x His-tag. Alternative name: TBEV NS1 protein. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In DPBS, pH 7. 4, 0.2µm filter sterilized. Source: Produced in HEK293 cells. Tick-borne encephalitis virus NS1 protein is fused at the C-terminus to a His-tag. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-4092