Learn More
Medchemexpress LLC HY-100489 500mg Medchemexpress, TBHQ CAS:1948-33-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100489500MG
Encompass_Preferred
Medchemexpress, HY-100489 500mg TBHQ CAS:1948-33-0 TBHQ (tert-Butylhydroquinone) is a widely used Nrf2 activator, protects against Doxorubicin (DOX)-induced cardiotoxicity through activation of Nrf2. TBHQ (tert-Butylhydroquinone) is also an ERK activator; rescues Dehydrocorydaline (DHC)-induced cell proliferation inhibitionin melanoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7735