Learn More
MSC Trinity Brand Industries Thickness Gage, 0.009 in., 304 Stainless Steel, 1/2 in. Width, 10 ft.
Supplier: MSC 01378926
This stainless steel thickness gage coil is designed for applications where resistance to corrosive environments and durability are essential. Manufactured from 300 series, full hard stainless steel, this 0.009 in. thick material offers outstanding wear resistance for precise gaging needs.
- Constructed from 304 stainless steel, featuring 18% chromium and 8% nickel austenitic alloy composition.
- Supplied in a 10 ft. long coil measuring 1/2 in. in width.
- Thicknesses are clearly electrochemically etched on the material with both decimal and metric equivalents.
- Engineered for performance in corrosive environments requiring high wear resistance.