Learn More
Medchemexpress LLC HY-100464 1mg , tBID CAS:1639895-85-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004641MG
Encompass_Preferred
HY-100464 1mg tBID CAS: 1639895-85-4 tBID is a selective inhibitor of homeodomain-interacting protein kinase 2 (HIPK2) with an IC50 of  0.33 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9859