Learn More
Accuflow Systems TBNG MFLEX FDA VITON #14 25FT
Supplier:  Accuflow Systems 9641214
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex FDA-Compliant Viton Precision Pump Tubing, L/S 14, 25'. Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work. Characte
Catalog No. 50-110-8821