Learn More
Accuflow Systems TBNG MFLEX FDA VITON #17 25FT
Supplier:  Accuflow Systems 9641217
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex FDA-Compliant Viton Precision Pump Tubing, L/S 17, 25' Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work. Character
Catalog No. 50-110-8924