Learn More
Accuflow Systems TBNG MFLEX SILICONE #15 250FT
Supplier:  Accuflow Systems 9640715
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex peroxide-cured silicone tubing, L/S 15, 250 ft. Cut to any length to fit your application Save money by buying in bulk—costs less to ship, too Reduce scrap—cut only as much as you need E
Catalog No. 50-110-8972