Learn More
Accuflow Systems TBNG MFLEX SILICONE #16 400FT
Supplier:  Accuflow Systems 9642316
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex BioPharm silicone tubing, L/S 16, 400 ft. Platinum-Cured silicone Ultra-smooth inner surface Fewer extractables Minimizes particle entrapment Perfect for lab, biotechnology, & pharmaceut
Catalog No. 50-110-8990
Only null left
Add to Cart
Request a Quote