Learn More
Accuflow Systems TBNG MFLEX TYGON E-LAB#25 50FT
Supplier:  Accuflow Systems 0650925
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Tygon E-Lab Tubing (E-3603), L/S 25, 50 ft. Specifications: Tubing formulation - Tygon E-Lab (E-3603) Pump Series - L/S Masterflex tubing size (L/S) - 25 Tubing ID (mm) - 4.8 Hose barb size (in.)
Catalog No. 50-110-8877