Learn More
Accuflow Systems TBNG MFLEX TYGON ELFL #24 25FT
Supplier:  Accuflow Systems 0644024
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon E-LFL Pump Tubing, L/S 24, 25 ft. High-performance pump tubing for challenging applications Nontoxic and nonhemolytic with excellent biocompatibility Longest life of all Tygon formulations
Catalog No. 50-110-8880