Learn More
Accuflow Systems TBNG MFLEX TYGON F&L #35 50FT
Supplier:  Accuflow Systems 0640135
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon Fuel & Lubricant tubing, L/S 35, 50 ft. Applications: Fuels and industrial lubricants—gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Characteristics: Resis
Catalog No. 50-110-8898