Learn More
Biocare Medical, LLC Tris Buffer Saline (TBS) Plus, 10X
SDP
Supplier:  Biocare Medical, LLC TBS942M
Encompass_Preferred
For laboratory use and procedures requiring a high quality Tris buffer saline. TBS is sodium azide and thimerosal free and provides superior pH stability. It can also be used for manual immunohistochemistry applications or on an automated staining system with the addition of surfactant.
Catalog No. NC0568555