Learn More
Alkali Scientific TBS / Tween 20 10X Solution, 1 L
SDP
Supplier:  Alkali Scientific TBS001CP
Encompass_Preferred
TBS / Tween 20 10X Solution is a powerful washing buffer used in a variety of biological and biochemical assays. This 1L solution is ideal for applications in immunohistochemistry, Western blotting, and enzyme-linked immunosorbent assays (ELISA). The formulation includes Tris, which maintains a stable pH, and Tween 20, a non-ionic detergent that helps reduce non-specific binding by blocking protein interaction sites. This buffer is essential for washing steps during immunodetection procedures, ensuring that only the specific antigen-antibody complex remains bound while unwanted proteins are removed. Its versatility makes it a staple in molecular biology, diagnostic laboratories, and research environments.
Catalog No. 50-006-61273