Learn More
Supply Solutions Tris Buffered Saline, with Tween 20, pH 8.0, 10 pack
Supplier:  Supply Solutions SIGMA T903910PAK
Encompass_Preferred
Tris Buffered Saline, with Tween 20, pH 8.0 has been used as a wash buffer in western blotting and enzyme linked immunosorbent assay.
Catalog No. NC0829162