Learn More
Abcam TC-2403 o x alate, alpha4beta2 nAChR selective agonist, 50MG
Supplier:  Abcam AB12055750MG
Encompass_Preferred
MW 252.27 g/mol, Purity >99%. High affinity α4β2 nAChR selective agonist (Ki = 26 nM). Selective for CNS nAChR over muscle, ganglionic or enteric ACh receptors (EC₅₀ = 732 nM in rat thalamic synaptosomes).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7490