Learn More
Medchemexpress LLC TC-2559 difumarate | 2454492-41-0 | 98.4% | 438.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1362071MG
Encompass_Preferred
TC-2559 difumarate is a central nervous system (CNS)-selective, orally active partial agonist of the α4β2 subtype of nicotinic acetylcholine receptors (nAChR).
  • Selectivity for α4β2 receptors over α2β4, α4β4, and α3β4 receptors, with EC50 values ranging from 10-30 μM.
  • Demonstrates an antinociceptive effect, reducing formalin-induced nociceptive responses in mice and inhibiting chronic constriction injury-induced neuropathic pain in rats.
  • Solubility of 43.84 mg/mL (99.99 mM) in H2O, requiring ultrasonic and warming for preparation.
Catalog No. 50-003-51452