Learn More
Medchemexpress LLC TC-A 2317 hydrochloride | 1245907-03-2 | MFCD19690936 | 99.6% | 392.93 g/mol | C19H29ClN6O | 25 MG

Supplier: Medchemexpress LLC HY10326625MG
TC-A 2317 hydrochloride is a research-grade Aurora A kinase inhibitor provided as the hydrochloride salt for in vitro and preclinical applications. It is a potent, selective inhibitor supplied as a solid with high reported purity.
- Potent Aurora A kinase inhibition (Ki = 1.2 nM).
- Selective over Aurora B kinase.
- High purity (~99.6%).
- Available in milligram quantities, including 25 MG.
- Solid physical form for standard laboratory storage and handling.
- Structural identifiers available (SMILES; molecular weight 392.93 g/mol).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.