Learn More
TARGETMOL CHEMICALS INC TC ASK 10 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T130995MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TC ASK 10 is a potent selective and orally active inhibitor of apoptosis signal-regulating kinase 1 (ASK1IC50 = 14 nM). The IC50 value for ASK2 is 0.51 uM. purity: 99%
Catalog No. 50-258-1383