Learn More
Medchemexpress LLC TC-G 1005 | 1415407-60-1 | 99.34% | 399.48 | 10 MG

Supplier: Medchemexpress LLC HY11017350MG
TC-G 1005 is a potent, selective, and orally active agonist of the BA receptor Takeda G protein-coupled receptor 5 (TGR5), with EC50s of 0.72 nM and 6.2 nM for hTGR5 and mTGR5, respectively. It can reduce glucose levels in vivo.
- Activates human and mouse TGR5 with EC50s of 0.72 nM and 6.2 nM, respectively.
- Stimulates GLP-1 secretion in ICR mice.
- Causes a 49% reduction in blood glucose AUC0-120 min in ICR mice.
- Significantly reduces blood glucose at 4, 6, 10, and 24 hours in db/db mice.
- Exhibits low plasma exposure in rats with a Cmax of 56 ng/mL and a t1/2 of 1.5 hours.
- Appearance: Solid.
- Color: Off-white to yellow.
- Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.