Learn More
Cole-Parmer Digi-Sense Type-K, 2-Wire Feedthrough, 1 / 2" NPT (M) Fitting, 48" PTFE Wire
Supplier:  Cole-Parmer 9387002
Encompass_Preferred
Use these two-wire feedthroughs to make connections inside vessels or chambers. Use with probes with miniconnectors. Feedthroughs withstand vacuum conditions or pressures to 100 psi. Stainless steel and epoxy construction. Temperature range: -40 to 250> F (-40 to 121> C). Feedthroughs have a 1/2'' NPT(M) fitting. Include 48''L, PTFE insulated, 24-gauge (0.0201'' wire dia) lead wires.
Catalog No. 50-196-1619