Learn More
Cole-Parmer Type-K Roller Probe; 10.5" L 4" L Bow Mini-Connector, Exposed 5ft Coil Cord
Supplier:  Cole-Parmer 0851695
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).
Catalog No. 50-196-1519