Learn More
Cole-Parmer Digi-Sense Type K Thermocouple Probe Insulated Wire Probe with Sealed Tip with Mini-Connector, 10ft L 24 Awg
Supplier:  Cole-Parmer 1852533
Encompass_Preferred
Do not subject type J exposed-junction probes to moisture as they will rust
Catalog No. 50-196-1547