Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TC-N 1752 | 1211866-85-1 | MFCD22123242 | >98.0% | 516.52 g·mol⁻¹ | C25H27F3N6O3 | 50 MG
SDP

Catalog No. 5000239915
Encompass_Preferred

A small-molecule research compound that functions as a potent, orally active inhibitor of Nav1.7 voltage-gated sodium channels. It shows activity across multiple Nav subtypes and has demonstrated analgesic efficacy in preclinical pain models. It is provided as a high-purity solid for use in in vitro and in vivo pharmacology and electrophysiology studies.

  • Potent Nav1.7 inhibition (hNav1.7 IC50 ≈ 0.17 μM).
  • Activity across multiple Nav channel subtypes.
  • Demonstrated analgesic efficacy in preclinical pain models.
  • High purity suitable for pharmacology and electrophysiology studies.
  • Soluble in DMSO for in vitro testing.

Catalog No. 50-002-39915 Supplier Medchemexpress LLC Supplier No. HY10740550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule research compound that functions as a potent, orally active inhibitor of Nav1.7 voltage-gated sodium channels. It shows activity across multiple Nav subtypes and has demonstrated analgesic efficacy in preclinical pain models. It is provided as a high-purity solid for use in in vitro and in vivo pharmacology and electrophysiology studies.

  • Potent Nav1.7 inhibition (hNav1.7 IC50 ≈ 0.17 μM).
  • Activity across multiple Nav channel subtypes.
  • Demonstrated analgesic efficacy in preclinical pain models.
  • High purity suitable for pharmacology and electrophysiology studies.
  • Soluble in DMSO for in vitro testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.