Learn More
Bellco Glass, Inc. TC Treated 6-Well Plate W/Lids
SDP
Supplier:  Bellco Glass, Inc. 130000309
Encompass_Preferred
6-well Plates are gamma-irradiated for sterility and available in both sterile and treated versions.Distinct alpha-numeric well designationsWell volume: 17.7 mLLid included with plateAvailable in individually packed units12-well, 24-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico. Style-TC Treated and Sterile, Lid-With Lid, Material-Polystyrene (PS), Color-Natural, Package Type-Individually Packed, Carton Qty-50
Catalog No. 50-233-0490