Learn More
Medchemexpress LLC TC11 | 100823-03-8 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129478100MG
Encompass_Preferred
TC11 is a phenylacetylamide derivative that functions as an MCL1 degrader and an activator of Caspase-9 and CDK1. It induces the degradation of MCL1, leading to apoptotic cell death during prolonged mitotic arrest.
  • Functions as an MCL1 degrader
  • Activates Caspase-9 and CDK1
  • Induces apoptotic cell death
  • Downregulates MCL1 expression
Catalog No. 50-003-40419