Learn More
Aviva Systems Biology Corp ELOA2 Antibody - middle region (ARP35916_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35916_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEB3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCEB3B. Alternate names: ELOA2, HsT832, MGC119351, TCEB3LApplicationWB. Species reactivity: Human
Catalog No. 50-014-55923