Learn More
Cayman Chemical TCEP-bIotIn 500ug
SDP
Supplier:  Cayman Chemical 25365500 UG
Encompass_Preferred
An affinity probe for protein lysine crotonylation; addition of the TCEP moiety to crotonylated lysine residues results in adduct formation that allows for detection or immobilization of crotonylated proteins through interaction with the biotin ligand; has been used in the detection and isolation of crotonylated histones in HEK293 cells
Catalog No. 50-293-3101