Learn More
AG Scientific Inc TCEP Hydrochloride, 1 G
Supplier:  AG Scientific Inc T11801GM
Encompass_Preferred
TCEP hydrochloride is a non-thiol reducing agent used to reduce and break disulfide bonds. TCEP HCl is widely preferred to the similar reducing agent DTT because of its greater effectiveness and resistance to oxidation from air exposure.CAS Number: 51805-45-9Molecular Weight: 286.6 DaChemical Formula: C9H15O6P HClSolubility: Soluble in waterStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7085