Learn More
Electron Microscopy Sciences (TCH), Thiocarbohydrazide, Highest Purity, 2231-57-4, 5g
SDP
Supplier:  Electron Microscopy Sciences 21900
Encompass_Preferred
Molecular Formula CH6N4S, Molecular Weight 106.152, b.p. 171°C (decomposed), TCH is a Osmiophilic reagent. It has an indefinite shelf life due to a special purification process.
Catalog No. 50-243-24