Learn More
Selleck Chemical LLC TCID
SDP
Supplier:  Selleck Chemical LLC S714010MG
Encompass_Preferred
TCID is a DUB inhibitor for ubiquitin C-terminal hydrolase L3 with IC50 of 0 6 M 125-fold selective to L1
Catalog No. 50-000-01104